missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Trimethylpyrazole 98.0+%, TCI America™

Catalog No. T27245G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T27245G 5 g
T272425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T27245G Supplier TCI America Supplier No. T27245G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1072-91-9
Molecular Formula C6H10N2
Molecular Weight (g/mol) 110.16
MDL Number MFCD00015536
InChI Key HNOQAFMOBRWDKQ-UHFFFAOYSA-N
Synonym 1,3,5-trimethyl-1h-pyrazole, 1h-pyrazole, 1,3,5-trimethyl, pyrazole, 1,3,5-trimethyl, akos pao-1133, pubchem20856, vitas-bb tbb000655, buttpark 15357-40, ksc490o8l, 1,3,5-trimethylpyrazole, 1,3,5-trimethyl-1h-pyrazole #
PubChem CID 14081
IUPAC Name 1,3,5-trimethylpyrazole
SMILES CC1=CC(=NN1C)C

Specifications

Melting Point 37°C
Color Yellow
Boiling Point 170°C
Quantity 5 g
UN Number 1325
Formula Weight 110.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 1,3,5-Trimethylpyrazole