accessibility menu, dialog, popup

UserName

CH3NNCH3H3C

CAS RN 1072-91-9

IUPAC Name: 1,3,5-trimethyl-1H-pyrazole
Synonyms: 1H-pyrazole, 1,3,5-trimethyl-
Molecular Weight (g/mol): 110.16
Molecular Formula: C6H10N2
InChi Key: HNOQAFMOBRWDKQ-UHFFFAOYSA-N
SMILES: CN1N=C(C)C=C1C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results