missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Trimethylcyclohexane 98.0+%, TCI America™

Catalog No. T066425ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T066425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T066425ML Supplier TCI America Supplier No. T066425ML

Chemical Identifiers

CAS 1839-63-0
Molecular Formula C9H18
Molecular Weight (g/mol) 126.24
MDL Number MFCD00042625
InChI Key ODNRTOSCFYDTKF-UHFFFAOYSA-N
PubChem CID 35364
IUPAC Name 1,3,5-trimethylcyclohexane
SMILES CC1CC(C)CC(C)C1

Specifications

Color Colorless
Boiling Point 140°C
Quantity 25 mL
UN Number 3295
Formula Weight 126.24
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3,5-Trimethylcyclohexane