accessibility menu, dialog, popup

UserName

CAS RN 1839-63-0

CH3CH3H3C

CAS RN 1839-63-0

IUPAC Name: 1,3,5-trimethylcyclohexane
Synonyms: 1,3,5-trimethylcyclohexane
Molecular Weight (g/mol): 126.24
Molecular Formula: C9H18
InChi Key: ODNRTOSCFYDTKF-UHFFFAOYSA-N
SMILES: CC1CC(C)CC(C)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results