missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Trimethylbenzene 97.0+%, TCI America™

Catalog No. T047025ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T047025ML 25 mL
T0470500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T047025ML Supplier TCI America Supplier No. T047025ML

Chemical Identifiers

CAS 108-67-8
Molecular Formula C9H12
Molecular Weight (g/mol) 120.195
MDL Number MFCD00008538
InChI Key AUHZEENZYGFFBQ-UHFFFAOYSA-N
Synonym mesitylene, sym-trimethylbenzene, benzene, 1,3,5-trimethyl, fleet-x, trimethylbenzol, 3,5-dimethyltoluene, s-trimethylbenzene, hsdb 92, 2,4,6-trimethylbenzene, 1,3,5-trimethyl-benzene
PubChem CID 7947
ChEBI CHEBI:34833
IUPAC Name 1,3,5-trimethylbenzene
SMILES CC1=CC(=CC(=C1)C)C

Specifications

Color Colorless
Boiling Point 164°C
Quantity 25 mL
UN Number 2325
Formula Weight 120.20
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3,5-Trimethylbenzene
EINECSNumber (3)-0007&(3)-3427
RTECSNumber OX6825000
TSCA Yes