missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Trimethoxybenzene, 99%

Catalog No. p-7021583
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1398109 10 g
AAA1398118 50 g
AAA1398130 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1398109 Supplier Thermo Scientific Chemicals Supplier No. A1398109
Only null left

CAS: 621-23-8 | C9H12O3 | 168.19 g/mol

1,3,5-Trimethoxybenzene is a key component of the Chinese rose odor. It is used as a cosmetic agents. It was employed as secondary standard in quantitative proton NMR spectroscopy of pharmaceuticals. It cleaves p-methoxybenzyl protecting group on various alcohols and acids

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3,5-Trimethoxybenzene is a key component of the Chinese rose odor. It is used as a cosmetic agents. It was employed as secondary standard in quantitative proton NMR spectroscopy of pharmaceuticals. It cleaves p-methoxybenzyl protecting group on various alcohols and acids

Solubility
Insoluble in water.

Notes
Store in a cool, dry, well-ventilated area. Store away from oxidizing agent.

Chemical Identifiers

CAS 621-23-8
Molecular Formula C9H12O3
Molecular Weight (g/mol) 168.19
MDL Number MFCD00008385
InChI Key LKUDPHPHKOZXCD-UHFFFAOYSA-N
Synonym phloroglucinol trimethyl ether, sym-trimethoxybenzene, benzene, 1,3,5-trimethoxy, 1,3,5-trimethyoxybenzene, unii-00vji3vg3d, 1,3,5-trimethoxy benzene, o,o,o-1,3,5-trimethylresorcinol, 00vji3vg3d, dsstox_cid_25963, dsstox_rid_81259
PubChem CID 69301
ChEBI CHEBI:31038
IUPAC Name 1,3,5-trimethoxybenzene
SMILES COC1=CC(OC)=CC(OC)=C1

Specifications

Melting Point 51°C to 53°C
Boiling Point 255°C
Flash Point 85°C (185°F)
Quantity 10 g
Beilstein 1307993
Solubility Information Insoluble in water.
Formula Weight 168.19
Percent Purity 99%
Chemical Name or Material 1,3,5-Trimethoxybenzene
Hazard Category H302
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264-P270-P301+P312-P330-P501c
EINECSNumber 210-673-4
RTECSNumber DC2810000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only