missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Trifluorobenzene 98.0+%, TCI America™

Catalog No. p-7136567
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
T10395G 5 g
T103925G 25 g
2 options

Catalog No. T10395G

Supplier: TCI America T10395G

Only null left

Chemical Identifiers

CAS 372-38-3
Molecular Formula C6H3F3
Molecular Weight (g/mol) 132.085
MDL Number MFCD00000333
InChI Key JXUKFFRPLNTYIV-UHFFFAOYSA-N
Synonym benzene, 1,3,5-trifluoro, sym-trifluorobenzene, unii-bn94c411f8, pubchem1069, 1,5-trifluorobenzene, acmc-209iru, 2,4,6-trifluorophenyl, benzene,3,5-trifluoro, 1,3,5-tri-fluorobenzene, 1,3,5-trifluoro-benzene
PubChem CID 9745
IUPAC Name 1,3,5-trifluorobenzene
SMILES C1=C(C=C(C=C1F)F)F

Specifications

Color Colorless
Boiling Point 76°C
Quantity 5 g
UN Number 1993
Formula Weight 132.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3,5-Trifluorobenzene