missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3,5-Trifluorobenzene, 98+%
Description
1,3,5-Trifluorobenzene was used in the synthesis of (-)-epicatechin and its 3-O-gallate. It is also used as an intermediate in organic chemical synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,3,5-Trifluorobenzene was used in the synthesis of (-)-epicatechin and its 3-O-gallate. It is also used as an intermediate in organic chemical synthesis.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
1,3,5-Trifluorobenzene was used in the synthesis of (-)-epicatechin and its 3-O-gallate. It is also used as an intermediate in organic chemical synthesis.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | -7°C to -4°C |
| Density | 1.279 |
| Boiling Point | 75°C to 76°C |
| Flash Point | −7°C (19°F) |
| Refractive Index | 1.415 |
| Quantity | 5 g |
| UN Number | UN2387 |
| Beilstein | 1906457 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 132.09 |
| Show More |
Safety and Handling
| Hazard Category | H225-H315-H319-H335 |
| Hazard Statement | GHS H Statement H225-H315-H319-H335 Highly flammable liquid and vapour. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLUOROBENZENE |
| EINECSNumber | 206-751-2 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only