missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Triethynylbenzene 98.0+%, TCI America™

Catalog No. T27605G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T27605G 5 g
T27601G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T27605G Supplier TCI America Supplier No. T27605G

Chemical Identifiers

CAS 7567-63-7
Molecular Formula C12H6
Molecular Weight (g/mol) 150.18
MDL Number MFCD00068621
InChI Key ZDRMMTYSQSIGRY-UHFFFAOYSA-N
PubChem CID 139048
IUPAC Name 1,3,5-triethynylbenzene
SMILES C#CC1=CC(=CC(=C1)C#C)C#C

Specifications

Melting Point 105°C
Quantity 5 g
Synonym benzene, 1,3,5-triethynyl, benzene,1,3,5-triethynyl, 1,3,5-phenylene-triacetylene, acmc-209ozh, 1,3,5-triethynyl-benzene, 1,3,5-triethynylbenzene, 1,3,5-triethynylbenzene 1g, benzene, 1,3,5-triethynyl-8ci, 9ci
Formula Weight 150.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3,5-Triethynylbenzene
TSCA No
Recommended Storage Refrigerator