accessibility menu, dialog, popup

UserName

CAS RN 7567-63-7

CHHCCH

CAS RN 7567-63-7

IUPAC Name: 1,3,5-triethynylbenzene
Synonyms: 1,3,5-triethynylbenzene
Molecular Weight (g/mol): 150.18
Molecular Formula: C12H6
InChi Key: ZDRMMTYSQSIGRY-UHFFFAOYSA-N
SMILES: C#CC1=CC(=CC(=C1)C#C)C#C
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)