missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3,5-Triethylbenzene 97.0+%, TCI America™
Supplier: TCI America T04265ML
Specifications
| 1,3,5-Triethylbenzene | |
| Colorless | |
| C12H18 | |
| 5 mL | |
| benzene, 1,3,5-triethyl, unii-02k328wwzq, 1,5-triethylbenzene, pubchem19794, benzene,3,5-triethyl, 1,3,5-triethyl benzene, ksc493q9l, wjympxjvhndzhd-uhfffaoysa, 1,3,5-triethylbenzene, 1,3,5-triethylbenzene, technical grade | |
| CCC1=CC(CC)=CC(CC)=C1 | |
| 162.28 | |
| 162.28 | |
| Liquid |
| 102-25-0 | |
| 218°C | |
| MFCD00009261 | |
| 3082 | |
| WJYMPXJVHNDZHD-UHFFFAOYSA-N | |
| 1,3,5-triethylbenzene | |
| 7602 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 102-25-0 | |
| 162.28 | |
| WJYMPXJVHNDZHD-UHFFFAOYSA-N | |
| 7602 | |
| CCC1=CC(CC)=CC(CC)=C1 |
| C12H18 | |
| MFCD00009261 | |
| benzene, 1,3,5-triethyl, unii-02k328wwzq, 1,5-triethylbenzene, pubchem19794, benzene,3,5-triethyl, 1,3,5-triethyl benzene, ksc493q9l, wjympxjvhndzhd-uhfffaoysa, 1,3,5-triethylbenzene, 1,3,5-triethylbenzene, technical grade | |
| 1,3,5-triethylbenzene |
Safety and Handling
EINECSNumber : (3)-3427
TSCA : Yes