missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Triethylbenzene 97.0+%, TCI America™

Catalog No. p-7136275
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mL
5 mL
Catalog No. Quantity
T04265ML 5 mL
T042625ML 25 mL
2 options

Catalog No. T04265ML

Supplier: TCI America T04265ML

Only null left

Chemical Identifiers

CAS 102-25-0
Molecular Formula C12H18
Molecular Weight (g/mol) 162.28
MDL Number MFCD00009261
InChI Key WJYMPXJVHNDZHD-UHFFFAOYSA-N
Synonym benzene, 1,3,5-triethyl, unii-02k328wwzq, 1,5-triethylbenzene, pubchem19794, benzene,3,5-triethyl, 1,3,5-triethyl benzene, ksc493q9l, wjympxjvhndzhd-uhfffaoysa, 1,3,5-triethylbenzene, 1,3,5-triethylbenzene, technical grade
PubChem CID 7602
IUPAC Name 1,3,5-triethylbenzene
SMILES CCC1=CC(CC)=CC(CC)=C1

Specifications

Color Colorless
Boiling Point 218°C
Quantity 5 mL
UN Number 3082
Formula Weight 162.28
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3,5-Triethylbenzene
EINECSNumber (3)-3427
TSCA Yes