missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3,5-Tribromobenzene 98.0+%, TCI America™
$29.83 - $447.38
Chemical Identifiers
| CAS | 626-39-1 |
|---|---|
| Molecular Formula | C6H3Br3 |
| Molecular Weight (g/mol) | 314.80 |
| MDL Number | MFCD00000080 |
| InChI Key | YWDUZLFWHVQCHY-UHFFFAOYSA-N |
| Synonym | benzene, 1,3,5-tribromo, 1,3,5-tribromo-benzene, unii-o3td0u1oaq, o3td0u1oaq, paragos 530416, 1,5-tribromobenzene, pubchem9590, 1,3,5-tribromobezene, benzene,3,5-tribromo, acmc-20a11o |
| PubChem CID | 12279 |
| IUPAC Name | 1,3,5-tribromobenzene |
| SMILES | BrC1=CC(Br)=CC(Br)=C1 |
Chemical Identifiers
| 626-39-1 | |
| 314.80 | |
| YWDUZLFWHVQCHY-UHFFFAOYSA-N | |
| 12279 | |
| BrC1=CC(Br)=CC(Br)=C1 |
| C6H3Br3 | |
| MFCD00000080 | |
| benzene, 1,3,5-tribromo, 1,3,5-tribromo-benzene, unii-o3td0u1oaq, o3td0u1oaq, paragos 530416, 1,5-tribromobenzene, pubchem9590, 1,3,5-tribromobezene, benzene,3,5-tribromo, acmc-20a11o | |
| 1,3,5-tribromobenzene |
Specifications
| 626-39-1 | |
| 271°C | |
| MFCD00000080 | |
| benzene, 1,3,5-tribromo, 1,3,5-tribromo-benzene, unii-o3td0u1oaq, o3td0u1oaq, paragos 530416, 1,5-tribromobenzene, pubchem9590, 1,3,5-tribromobezene, benzene,3,5-tribromo, acmc-20a11o | |
| BrC1=CC(Br)=CC(Br)=C1 | |
| 314.80 | |
| 314.80 | |
| Crystalline Powder |
| 122°C | |
| C6H3Br3 | |
| 5 g | |
| YWDUZLFWHVQCHY-UHFFFAOYSA-N | |
| 1,3,5-tribromobenzene | |
| 12279 | |
| ≥98.0% (GC) | |
| 1,3,5-Tribromobenzene |
Safety and Handling
RTECSNumber : DC1957200
TSCA : Yes