missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Triazine, 97%

CAS: 290-87-9 | C3H3N3 | 81.08 g/mol

Supplier:  Thermo Scientific Chemicals L1691114

 View more versions of this product

Catalog No. AAL1691114


Only null left
Explore available promotions

Description

Description

1,3,5-Triazine is a useful replacement for HCN in the Gattermann reaction for the synthesis of aromatic aldehydes. It reacts with nucleophiles, e.g. amines, which is utilized in quinazoline synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3,5-Triazine is a useful replacement for HCN in the Gattermann reaction for the synthesis of aromatic aldehydes. It reacts with nucleophiles, e.g. amines, which is utilized in quinazoline synthesis.

Solubility
Soluble in ethanol (50 mg/ml), and methanol (100 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is hygroscopic in nature. Store under dry inert gas.
Specifications

Specifications

1,3,5-Triazine
77°C to 83°C (decomposition)
114°C
MFCD00006044
104790
s-triazine, sym-triazine, cyanidine, vedita 250, unii-8b5f4cm81e, vedita, 1,5-triazine, hydrogen cyanide trimer, acmc-209h6t
JIHQDMXYYFUGFV-UHFFFAOYSA-N
1,3,5-triazine
9262
81.08
290-87-9
1.38
C3H3N3
25 g
Moisture sensitive
Soluble in ethanol (50mg/ml),and methanol (100mg/ml).
C1=NC=NC=N1
81.08
CHEBI:30259
97%

Chemical Identifiers

290-87-9
81.08
JIHQDMXYYFUGFV-UHFFFAOYSA-N
9262
1,3,5-triazine
C3H3N3
MFCD00006044
s-triazine, sym-triazine, cyanidine, vedita 250, unii-8b5f4cm81e, vedita, 1,5-triazine, hydrogen cyanide trimer, acmc-209h6t
CHEBI:30259
C1=NC=NC=N1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c

H302-H315-H318-H335

EINECSNumber : 206-028-1

RTECSNumber : XY2957000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only