missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Triazine, 97%

Catalog No. p-7046949
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1691103 1 g
AAL1691106 5 g
AAL1691114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAL1691103 Supplier Thermo Scientific Chemicals Supplier No. L1691103
Only null left
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 290-87-9 | C3H3N3 | 81.08 g/mol

1,3,5-Triazine is a useful replacement for HCN in the Gattermann reaction for the synthesis of aromatic aldehydes. It reacts with nucleophiles, e.g. amines, which is utilized in quinazoline synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3,5-Triazine is a useful replacement for HCN in the Gattermann reaction for the synthesis of aromatic aldehydes. It reacts with nucleophiles, e.g. amines, which is utilized in quinazoline synthesis.

Solubility
Soluble in ethanol (50 mg/ml), and methanol (100 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is hygroscopic in nature. Store under dry inert gas.

Chemical Identifiers

CAS 290-87-9
Molecular Formula C3H3N3
Molecular Weight (g/mol) 81.08
MDL Number MFCD00006044
InChI Key JIHQDMXYYFUGFV-UHFFFAOYSA-N
Synonym s-triazine, sym-triazine, cyanidine, vedita 250, unii-8b5f4cm81e, vedita, 1,5-triazine, hydrogen cyanide trimer, acmc-209h6t
PubChem CID 9262
ChEBI CHEBI:30259
IUPAC Name 1,3,5-triazine
SMILES C1=NC=NC=N1

Specifications

Melting Point 77°C to 83°C (decomposition)
Density 1.38
Boiling Point 114°C
Quantity 1 g
Beilstein 104790
Sensitivity Moisture sensitive
Solubility Information Soluble in ethanol (50mg/ml),and methanol (100mg/ml).
Formula Weight 81.08
Percent Purity 97%
Chemical Name or Material 1,3,5-Triazine
Hazard Category H302-H315-H318-H335
Hazard Statement GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
EINECSNumber 206-028-1
RTECSNumber XY2957000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only