Learn More
1,3,5-Triazine, 97%
CAS: 290-87-9 | C3H3N3 | 81.08 g/mol
Supplier: Thermo Scientific Chemicals L1691106
Description
1,3,5-Triazine is a useful replacement for HCN in the Gattermann reaction for the synthesis of aromatic aldehydes. It reacts with nucleophiles, e.g. amines, which is utilized in quinazoline synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,3,5-Triazine is a useful replacement for HCN in the Gattermann reaction for the synthesis of aromatic aldehydes. It reacts with nucleophiles, e.g. amines, which is utilized in quinazoline synthesis.
Solubility
Soluble in ethanol (50 mg/ml), and methanol (100 mg/ml).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is hygroscopic in nature. Store under dry inert gas.
Specifications
| 1,3,5-Triazine | |
| 77°C to 83°C (decomposition) | |
| 114°C | |
| MFCD00006044 | |
| 104790 | |
| s-triazine, sym-triazine, cyanidine, vedita 250, unii-8b5f4cm81e, vedita, 1,5-triazine, hydrogen cyanide trimer, acmc-209h6t | |
| JIHQDMXYYFUGFV-UHFFFAOYSA-N | |
| 1,3,5-triazine | |
| 9262 | |
| 81.08 |
| 290-87-9 | |
| 1.38 | |
| C3H3N3 | |
| 5 g | |
| Moisture sensitive | |
| Soluble in ethanol (50mg/ml),and methanol (100mg/ml). | |
| C1=NC=NC=N1 | |
| 81.08 | |
| CHEBI:30259 | |
| 97% |
Chemical Identifiers
| 290-87-9 | |
| 81.08 | |
| JIHQDMXYYFUGFV-UHFFFAOYSA-N | |
| 9262 | |
| 1,3,5-triazine |
| C3H3N3 | |
| MFCD00006044 | |
| s-triazine, sym-triazine, cyanidine, vedita 250, unii-8b5f4cm81e, vedita, 1,5-triazine, hydrogen cyanide trimer, acmc-209h6t | |
| CHEBI:30259 | |
| C1=NC=NC=N1 |
Safety and Handling
GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
H302-H315-H318-H335
EINECSNumber : 206-028-1
RTECSNumber : XY2957000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only