missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Tri(p-tolyl)benzene 96.0+%, TCI America™

Catalog No. T33705G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T33705G 5 g
T33701G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T33705G Supplier TCI America Supplier No. T33705G
Only null left

Chemical Identifiers

CAS 50446-43-0
Molecular Formula C27H24
Molecular Weight (g/mol) 348.49
MDL Number MFCD00192798
InChI Key XMGRUKCVUYLTKU-UHFFFAOYSA-N
Synonym 1,3,5-Tris(4-Methylphenyl)benzene
PubChem CID 504157
IUPAC Name 4'-methyl-3,5-bis(4-methylphenyl)-1,1'-biphenyl
SMILES CC1=CC=C(C=C1)C1=CC(=CC(=C1)C1=CC=C(C)C=C1)C1=CC=C(C)C=C1

Specifications

Melting Point 176°C
Color White-Yellow
Quantity 5 g
Formula Weight 348.49
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3,5-Tri(p-tolyl)benzene