accessibility menu, dialog, popup

UserName

CH3H3CCH3

CAS RN 50446-43-0

IUPAC Name: 4'-methyl-3,5-bis(4-methylphenyl)-1,1'-biphenyl
Synonyms: 4'-methyl-3,5-bis(4-methylphenyl)-1,1'-biphenyl
Molecular Weight (g/mol): 348.49
Molecular Formula: C27H24
InChi Key: XMGRUKCVUYLTKU-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C1=CC(=CC(=C1)C1=CC=C(C)C=C1)C1=CC=C(C)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results