Learn More
Thermo Scientific Chemicals 1,3,5-Tri-O-benzoyl-2-deoxy-2-fluoro-alpha-D-arabinofuranose
CAS: 97614-43-2 | C26H21FO7 | 464.445 g/mol
Supplier: Thermo Scientific Chemicals J6680506
Description
It is a pharmaceutical intermediate and is used as an OLED materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is a pharmaceutical intermediate and is used as an OLED materials.
Solubility
Soluble in chloroform or dichloromethane
Notes
Refrigerate. Keep away from heat, oxidizing agents.
Specifications
| 1,3,5-Tri-O-benzoyl-2-deoxy-2-fluoro-alpha-D-arabinofuranose | |
| 76°C to 78°C | |
| C26H21FO7 | |
| 5 g | |
| Soluble in chloroform or dichloromethane | |
| C1=CC=C(C=C1)C(=O)OCC2C(C(C(O2)OC(=O)C3=CC=CC=C3)F)OC(=O)C4=CC=CC=C4 | |
| 464.445 | |
| 464.44 |
| 97614-43-2 | |
| White | |
| MFCD00083339 | |
| 2-deoxy-2-fluoro-1,3,5-tri-o-benzoyl-alpha-d-arabinofuranose, 2-deoxy-2-fluoro-1,3,5-tri-o-benzoyl-alpha-d-ribofuranose, 2-deoxy-2-fluoro-1,3,5-tri-o-benzoyl-, 2-deoxy-2-fluoro-1,3,5-tri-o-benzoyl-, 1,3,5-tri-o-benzoyl-2-deoxy-2-fluoro-a-d-arabinofuranose, 2r,3r,4s,5r-3,5-bis benzoyloxy-4-fluorooxolan-2-yl methyl benzoate, 2r,3r,4s,5r-3,5-bis benzoyloxy-4-fluorooxolan-2-yl methyl benzoate, 2r,3s,4r,5r-5-benzoyloxymethyl-3-fluoro-tetrahydrofuran-2,4-diyl dibenzoate, 2-deoxy-2-fluoro-1,3,5-tri-o-benzoyl-d-ribofuranose, 2-deoxy-2-fluoro-1,3,5-tri-o-benzoyl-d-ribofuranose, pubchem20401 | |
| JOAHVPNLVYCSAN-UXGLMHHASA-N | |
| [(2R,3R,4S,5R)-3,5-dibenzoyloxy-4-fluorooxolan-2-yl]methyl benzoate | |
| 11754171 | |
| Powder |
Chemical Identifiers
| 97614-43-2 | |
| 464.445 | |
| JOAHVPNLVYCSAN-UXGLMHHASA-N | |
| 11754171 | |
| C1=CC=C(C=C1)C(=O)OCC2C(C(C(O2)OC(=O)C3=CC=CC=C3)F)OC(=O)C4=CC=CC=C4 |
| C26H21FO7 | |
| MFCD00083339 | |
| 2-deoxy-2-fluoro-1,3,5-tri-o-benzoyl-alpha-d-arabinofuranose, 2-deoxy-2-fluoro-1,3,5-tri-o-benzoyl-alpha-d-ribofuranose, 2-deoxy-2-fluoro-1,3,5-tri-o-benzoyl-, 2-deoxy-2-fluoro-1,3,5-tri-o-benzoyl-, 1,3,5-tri-o-benzoyl-2-deoxy-2-fluoro-a-d-arabinofuranose, 2r,3r,4s,5r-3,5-bis benzoyloxy-4-fluorooxolan-2-yl methyl benzoate, 2r,3r,4s,5r-3,5-bis benzoyloxy-4-fluorooxolan-2-yl methyl benzoate, 2r,3s,4r,5r-5-benzoyloxymethyl-3-fluoro-tetrahydrofuran-2,4-diyl dibenzoate, 2-deoxy-2-fluoro-1,3,5-tri-o-benzoyl-d-ribofuranose, 2-deoxy-2-fluoro-1,3,5-tri-o-benzoyl-d-ribofuranose, pubchem20401 | |
| [(2R,3R,4S,5R)-3,5-dibenzoyloxy-4-fluorooxolan-2-yl]methyl benzoate |
Safety and Handling
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only