missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 1,3,5-Tri-O-benzoyl-2-deoxy-2-fluoro-alpha-D-arabinofuranose

Catalog No. p-7043275
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6680503 1 g
AAJ6680506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6680503 Supplier Thermo Scientific Chemicals Supplier No. J6680503

CAS: 97614-43-2 | C26H21FO7 | 464.445 g/mol

It is a pharmaceutical intermediate and is used as an OLED materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a pharmaceutical intermediate and is used as an OLED materials.

Solubility
Soluble in chloroform or dichloromethane

Notes
Refrigerate. Keep away from heat, oxidizing agents.

Chemical Identifiers

CAS 97614-43-2
Molecular Formula C26H21FO7
Molecular Weight (g/mol) 464.445
MDL Number MFCD00083339
InChI Key JOAHVPNLVYCSAN-UXGLMHHASA-N
Synonym 2-deoxy-2-fluoro-1,3,5-tri-o-benzoyl-alpha-d-arabinofuranose, 2r,3s,4r,5r-5-benzoyloxy methyl-3-fluorotetrahydrofuran-2,4-diyl dibenzoate, 2-deoxy-2-fluoro-1,3,5-tri-o-benzoyl-, a-d-arabinofuranose, 1,3,5-tri-o-benzoyl-2-deoxy-2-fluoro-a-d-arabinofuranose, 2r,3r,4s,5r-3,5-bis benzoyloxy-4-fluorooxolan-2-yl methyl benzoate, 2r,3s,4r,5r-5-benzoyloxymethyl-3-fluoro-tetrahydrofuran-2,4-diyl dibenzoate, 1,3,5-tri-o-benzoyl-2-deoxy-2-fluoro-md-arabinose, 2-deoxy-2-fluoro-1,3,5-tri-o-benzoyl-d-ribofuranose, pubchem20401, 2-deoxy-2-fluoro-1,3,5-tri-o-benzoyl-alpha-d-ribofuranose
PubChem CID 11754171
IUPAC Name [(2R,3R,4S,5R)-3,5-dibenzoyloxy-4-fluorooxolan-2-yl]methyl benzoate
SMILES C1=CC=C(C=C1)C(=O)OCC2C(C(C(O2)OC(=O)C3=CC=CC=C3)F)OC(=O)C4=CC=CC=C4

Specifications

Melting Point 76°C to 78°C
Color White
Quantity 1 g
Solubility Information Soluble in chloroform or dichloromethane
Formula Weight 464.44
Physical Form Powder
Chemical Name or Material 1,3,5-Tri-O-benzoyl-2-deoxy-2-fluoro-alpha-D-arabinofuranose
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only