missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3,5-Tri(1-naphthyl)benzene 98.0+%, TCI America™
Supplier: TCI America T31931G
Specifications
| 1,3,5-Tri(1-naphthyl)benzene | |
| 186°C | |
| 1 g | |
| C1=CC=C2C(=C1)C=CC=C2C3=CC(=CC(=C3)C4=CC=CC5=CC=CC=C54)C6=CC=CC7=CC=CC=C76 | |
| 456.588 | |
| 456.59 | |
| Crystalline Powder |
| 7059-70-3 | |
| C36H24 | |
| ZVUZLHDATWCFQW-UHFFFAOYSA-N | |
| 1-(3,5-dinaphthalen-1-ylphenyl)naphthalene | |
| 15208120 | |
| ≥98.0% (HPLC) |
Chemical Identifiers
| 7059-70-3 | |
| 456.588 | |
| 15208120 | |
| C1=CC=C2C(=C1)C=CC=C2C3=CC(=CC(=C3)C4=CC=CC5=CC=CC=C54)C6=CC=CC7=CC=CC=C76 |
| C36H24 | |
| ZVUZLHDATWCFQW-UHFFFAOYSA-N | |
| 1-(3,5-dinaphthalen-1-ylphenyl)naphthalene |
Safety and Handling
TSCA : No