missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Tri(1-naphthyl)benzene 98.0+%, TCI America™

Catalog No. p-7137632
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3193200MG 200 mg
T31931G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T3193200MG Supplier TCI America Supplier No. T3193200MG

Chemical Identifiers

CAS 7059-70-3
Molecular Formula C36H24
Molecular Weight (g/mol) 456.588
InChI Key ZVUZLHDATWCFQW-UHFFFAOYSA-N
PubChem CID 15208120
IUPAC Name 1-(3,5-dinaphthalen-1-ylphenyl)naphthalene
SMILES C1=CC=C2C(=C1)C=CC=C2C3=CC(=CC(=C3)C4=CC=CC5=CC=CC=C54)C6=CC=CC7=CC=CC=C76

Specifications

Melting Point 186°C
Quantity 200 mg
Formula Weight 456.59
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3,5-Tri(1-naphthyl)benzene