missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Pentanetricarbonitrile 96.0+%, TCI America™

Catalog No. P1631500MG
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1631500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P1631500MG Supplier TCI America Supplier No. P1631500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4379-04-8
Molecular Formula C8H9N3
Molecular Weight (g/mol) 147.18
InChI Key RXIMZKYZCDNHPG-UHFFFAOYSA-N
Synonym 4-Cyanoheptanedinitrile
PubChem CID 252838
IUPAC Name pentane-1,3,5-tricarbonitrile
SMILES C(CC(CCC#N)C#N)C#N

Specifications

Boiling Point 190°C
Quantity 500 mg
UN Number 3276
Formula Weight 147.18
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3,5-Pentanetricarbonitrile