accessibility menu, dialog, popup

UserName

NNN

CAS RN 4379-04-8

IUPAC Name: pentane-1,3,5-tricarbonitrile
Synonyms: pentane-1,3,5-tricarbonitrile
Molecular Weight (g/mol): 147.18
Molecular Formula: C8H9N3
InChi Key: RXIMZKYZCDNHPG-UHFFFAOYSA-N
SMILES: N#CCCC(CCC#N)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results