missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Cyclohexanetriol (cis- and trans- mixture) 95.0+%, TCI America™

Catalog No. p-7124761
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C10515G 5 g
C105125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C10515G Supplier TCI America Supplier No. C10515G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2041-15-8
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.16
MDL Number MFCD00043062,MFCD00070580
InChI Key FSDSKERRNURGGO-UHFFFAOYSA-N
PubChem CID 230351
IUPAC Name cyclohexane-1,3,5-triol
SMILES OC1CC(O)CC(O)C1

Specifications

Color White
Quantity 5 g
Formula Weight 132.16
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3,5-Cyclohexanetriol (cis- and trans- mixture)