missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Benzenetrimethanol 95.0+%, TCI America™

Catalog No. p-7123029
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B35861G 1 g
B35865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B35861G Supplier TCI America Supplier No. B35861G

Chemical Identifiers

CAS 4464-18-0
Molecular Formula C9H12O3
Molecular Weight (g/mol) 168.192
MDL Number MFCD01108484
InChI Key SQAMZFDWYRVIMG-UHFFFAOYSA-N
Synonym 1,3,5-Tris(hydroxymethyl)benzene
PubChem CID 2748048
IUPAC Name [3,5-bis(hydroxymethyl)phenyl]methanol
SMILES C1=C(C=C(C=C1CO)CO)CO

Specifications

Melting Point 78°C
Color White-Yellow
Quantity 1 g
Formula Weight 168.19
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3,5-Benzenetrimethanol