missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3,5-Benzenetricarboxylic Acid 98.0+%, TCI America™
$57.75 - $419.69
Chemical Identifiers
| CAS | 554-95-0 |
|---|---|
| Molecular Formula | C9H6O6 |
| Molecular Weight (g/mol) | 210.14 |
| MDL Number | MFCD00002517 |
| InChI Key | QMKYBPDZANOJGF-UHFFFAOYSA-N |
| Synonym | trimesic acid, 1,3,5-benzenetricarboxylic acid, trimesinic acid, trimesitinic acid, 5-carboxyisophthalic acid, 1,3,5-benzene tricarboxylic acid, 1,3,5-tricarboxybenzene, unii-ou36oo5mtn, ou36oo5mtn, chembl77562 |
| PubChem CID | 11138 |
| ChEBI | CHEBI:46032 |
| IUPAC Name | benzene-1,3,5-tricarboxylic acid |
| SMILES | OC(=O)C1=CC(=CC(=C1)C(O)=O)C(O)=O |
Chemical Identifiers
| 554-95-0 | |
| 210.14 | |
| QMKYBPDZANOJGF-UHFFFAOYSA-N | |
| 11138 | |
| benzene-1,3,5-tricarboxylic acid |
| C9H6O6 | |
| MFCD00002517 | |
| trimesic acid, 1,3,5-benzenetricarboxylic acid, trimesinic acid, trimesitinic acid, 5-carboxyisophthalic acid, 1,3,5-benzene tricarboxylic acid, 1,3,5-tricarboxybenzene, unii-ou36oo5mtn, ou36oo5mtn, chembl77562 | |
| CHEBI:46032 | |
| OC(=O)C1=CC(=CC(=C1)C(O)=O)C(O)=O |
Specifications
| 554-95-0 | |
| C9H6O6 | |
| 25 g | |
| QMKYBPDZANOJGF-UHFFFAOYSA-N | |
| benzene-1,3,5-tricarboxylic acid | |
| 11138 | |
| 210.14 | |
| Crystalline Powder |
| White-Yellow | |
| MFCD00002517 | |
| trimesic acid, 1,3,5-benzenetricarboxylic acid, trimesinic acid, trimesitinic acid, 5-carboxyisophthalic acid, 1,3,5-benzene tricarboxylic acid, 1,3,5-tricarboxybenzene, unii-ou36oo5mtn, ou36oo5mtn, chembl77562 | |
| OC(=O)C1=CC(=CC(=C1)C(O)=O)C(O)=O | |
| 210.14 | |
| CHEBI:46032 | |
| ≥98.0% (T) | |
| 1,3,5-Benzenetricarboxylic Acid |
Safety and Handling
EINECSNumber : (3)-1279
TSCA : Yes