missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Benzenetricarboxylic acid, 98%

Catalog No. p-3777359
Change view
Click to view available options
Quantity:
50 g
Packaging:
Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC105350500 50 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC105350500 Supplier Thermo Scientific Chemicals Supplier No. 105350500
Only null left

1, 3, 5-Benzenetricarboxylic acid, 98%, C9H6O6, CAS Number-554-95-0, 1,3,5-benzene tricarboxylic acid, 1,3,5-benzenetricarboxylic acid, 1,3,5-tricarboxybenzene, 5-carboxyisophthalic acid, chembl77562, ou36oo5mtn, trimesic acid, trimesinic acid, trimesitinic acid, unii-ou36oo5mtn, 500g | CAS: 554-95-0 | C9H6O6 | 210.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 554-95-0
Molecular Formula C9H6O6
Molecular Weight (g/mol) 210.14
MDL Number MFCD00002517
InChI Key QMKYBPDZANOJGF-UHFFFAOYSA-N
Synonym trimesic acid, 1,3,5-benzenetricarboxylic acid, trimesinic acid, trimesitinic acid, 5-carboxyisophthalic acid, 1,3,5-benzene tricarboxylic acid, 1,3,5-tricarboxybenzene, unii-ou36oo5mtn, ou36oo5mtn, chembl77562
PubChem CID 11138
ChEBI CHEBI:46032
IUPAC Name benzene-1,3,5-tricarboxylic acid
SMILES OC(=O)C1=CC(=CC(=C1)C(O)=O)C(O)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 380.0°C
Flash Point >250°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Plastic bottle
Linear Formula C6H3(CO2H)3
Quantity 50 g
Beilstein 09, 978
Solubility Information Solubility in water: 2.6g/L (25°C)- 7.9g/L (50°C). Other solubilities: 1.24g/100g ethyl ether- 8g/100g methanol,0.46g/100g acetic acid- >20.00g/100g thf (25°C),1.88g/100g acetone- <0.01g/100g xylene (25°C),<0.01g/1
Formula Weight 210.14
Percent Purity 98%
Chemical Name or Material 1, 3, 5-Benzenetricarboxylic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and
EINECSNumber 209-077-7

RUO – Research Use Only