missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3,5-Benzenetricarbonyl Trichloride 98.0+%, TCI America™
Supplier: TCI America T1262250G
Specifications
| 1,3,5-Benzenetricarbonyl Trichloride | |
| White-Yellow | |
| C9H3Cl3O3 | |
| 250 g | |
| trimesoyl chloride, 1,3,5-benzenetricarbonyl trichloride, 1,3,5-benzenetricarboxylic acid chloride, 1,3,5-benzenetricarbonyl chloride, trimesic acid trichloride, 1,3,5-benzenetricarboxylic chloride, benzene-1,3,5-tricarbonyl trichloride, unii-5866peo79w, 1,5-benzenetricarbonyl trichloride, 1,3,5-benzenetricarboxylicacidchloride | |
| ClC(=O)C1=CC(=CC(=C1)C(Cl)=O)C(Cl)=O | |
| 265.47 | |
| 265.47 | |
| Crystalline Lumps |
| 4422-95-1 | |
| 181°C | |
| MFCD00000679 | |
| 3261 | |
| UWCPYKQBIPYOLX-UHFFFAOYSA-N | |
| benzene-1,3,5-tricarbonyl trichloride | |
| 78138 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 4422-95-1 | |
| 265.47 | |
| UWCPYKQBIPYOLX-UHFFFAOYSA-N | |
| 78138 | |
| ClC(=O)C1=CC(=CC(=C1)C(Cl)=O)C(Cl)=O |
| C9H3Cl3O3 | |
| MFCD00000679 | |
| trimesoyl chloride, 1,3,5-benzenetricarbonyl trichloride, 1,3,5-benzenetricarboxylic acid chloride, 1,3,5-benzenetricarbonyl chloride, trimesic acid trichloride, 1,3,5-benzenetricarboxylic chloride, benzene-1,3,5-tricarbonyl trichloride, unii-5866peo79w, 1,5-benzenetricarbonyl trichloride, 1,3,5-benzenetricarboxylicacidchloride | |
| benzene-1,3,5-tricarbonyl trichloride |
Safety and Handling
EINECSNumber : (3)-4605
TSCA : Yes