missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Benzenetricarbonyl Trichloride 98.0+%, TCI America™

Catalog No. p-7136687
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T126225G 25 g
T1262250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T126225G Supplier TCI America Supplier No. T126225G

Chemical Identifiers

CAS 4422-95-1
Molecular Formula C9H3Cl3O3
Molecular Weight (g/mol) 265.47
MDL Number MFCD00000679
InChI Key UWCPYKQBIPYOLX-UHFFFAOYSA-N
Synonym trimesoyl chloride, 1,3,5-benzenetricarbonyl trichloride, 1,3,5-benzenetricarboxylic acid chloride, 1,3,5-benzenetricarbonyl chloride, trimesic acid trichloride, 1,3,5-benzenetricarboxylic chloride, benzene-1,3,5-tricarbonyl trichloride, unii-5866peo79w, 1,5-benzenetricarbonyl trichloride, 1,3,5-benzenetricarboxylicacidchloride
PubChem CID 78138
IUPAC Name benzene-1,3,5-tricarbonyl trichloride
SMILES ClC(=O)C1=CC(=CC(=C1)C(Cl)=O)C(Cl)=O

Specifications

Color White-Yellow
Boiling Point 181°C
Quantity 25 g
UN Number 3261
Formula Weight 265.47
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 1,3,5-Benzenetricarbonyl Trichloride
EINECSNumber (3)-4605
TSCA Yes