missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Benzenetricarbonitrile 98.0+%, TCI America™

Catalog No. b42941g
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B42941G 1 g
B42945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B42941G Supplier TCI America Supplier No. B42941G
Only null left

Chemical Identifiers

CAS 10365-94-3
Molecular Formula C9H3N3
Molecular Weight (g/mol) 153.144
MDL Number MFCD00019746
InChI Key SGLGUTWNGVJXPP-UHFFFAOYSA-N
Synonym 1,3,5-Tricyanobenzene
PubChem CID 82587
IUPAC Name benzene-1,3,5-tricarbonitrile
SMILES C1=C(C=C(C=C1C#N)C#N)C#N

Specifications

Melting Point 259°C
Color White-Yellow
Quantity 1 g
UN Number 3439
Formula Weight 153.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3,5-Benzenetricarbonitrile
RTECSNumber DC1958000
TSCA No