accessibility menu, dialog, popup

UserName

NNN

CAS RN 10365-94-3

IUPAC Name: benzene-1,3,5-tricarbonitrile
Synonyms: benzene-1,3,5-tricarbonitrile
Molecular Weight (g/mol): 153.14
Molecular Formula: C9H3N3
InChi Key: SGLGUTWNGVJXPP-UHFFFAOYSA-N
SMILES: N#CC1=CC(=CC(=C1)C#N)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results