missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Adamantanetriol 97.0+%, TCI America™

Catalog No. p-7117578
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1682250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A1682250MG Supplier TCI America Supplier No. A1682250MG

Chemical Identifiers

CAS 99181-50-7
Molecular Formula C10H16O3
Molecular Weight (g/mol) 184.235
InChI Key MCYBYTIPMYLHAK-UHFFFAOYSA-N
Synonym 1,3,5-Trihydroxyadamantane
PubChem CID 9899022
IUPAC Name adamantane-1,3,5-triol
SMILES C1C2CC3(CC1(CC(C2)(C3)O)O)O

Specifications

Color White
Quantity 250 mg
Formula Weight 184.24
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3,5-Adamantanetriol