accessibility menu, dialog, popup

UserName

OHHOOH

CAS RN 99181-50-7

IUPAC Name: adamantane-1,3,5-triol
Synonyms: adamantane-1,3,5-triol
Molecular Weight (g/mol): 184.24
Molecular Formula: C10H16O3
InChi Key: MCYBYTIPMYLHAK-UHFFFAOYSA-N
SMILES: OC12CC3CC(O)(C1)CC(O)(C3)C2
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results