missing translation for 'onlineSavingsMsg'
Learn More

1,3,4-Triphenyl-4,5-dihydro-1H-1,2,4-triazol-5-ylidene

Catalog No. AC363940010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC363940010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC363940010 Supplier Thermo Scientific Chemicals Supplier No. 363940010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 166773-08-6 | C20H15N3 | 297.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent

Chemical Identifiers

CAS 166773-08-6
Molecular Formula C20H15N3
Molecular Weight (g/mol) 297.36
MDL Number MFCD00801868
InChI Key QZHWOLKBXYORRO-UHFFFAOYSA-N
Synonym 2,4,5-triphenyl-4h-1,2,4-triazol-2-ium-3-ide, 1,3,4-triphenyl-4,5-dihydro-1h-1,2,4-triazol-5-ylidene, 1?3?4-triphenyl-4?5-dihydro-1h-1?2?4-triazol-5-ylidene
PubChem CID 2733186
SMILES C1=CC=C(C=C1)C2=N[N+](=[C-]N2C3=CC=CC=C3)C4=CC=CC=C4

Specifications

Melting Point 150°C
Color Beige
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 1 g
Formula Weight 297.36
Physical Form Crystalline Powder
Chemical Name or Material 1,3,4-Triphenyl-4,5-dihydro-1H-1,2,4-triazol-5-ylidene

RUO – Research Use Only