CAS RN 166773-08-6
IUPAC Name:
1,3,4-triphenyl-4,5-dihydro-1H-1,2,4-triazole
Synonyms:
1,3,4-triphenyl-5H-1,2,4-triazole
Molecular Weight (g/mol):
299.38
Molecular Formula:
C20H17N3
InChi Key:
NNVGTIRYCCWRSL-UHFFFAOYSA-N
SMILES:
C1N(N=C(N1C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
Promotions Available
1,3,4-Triphenyl-4,5-dihydro-1H-1,2,4-triazol-5-ylidene
1,3,4-Triphenyl-4,5-dihydro-1H-1,2,4-triazol-5-ylidene
CAS: 166773-08-6 Molecular Formula: C20H15N3 Molecular Weight (g/mol): 297.36 MDL Number: MFCD00801868 InChI Key: QZHWOLKBXYORRO-UHFFFAOYSA-N Synonym: 2,4,5-triphenyl-4h-1,2,4-triazol-2-ium-3-ide,1,3,4-triphenyl-4,5-dihydro-1h-1,2,4-triazol-5-ylidene,1?3?4-triphenyl-4?5-dihydro-1h-1?2?4-triazol-5-ylidene PubChem CID: 2733186 SMILES: C1=CC=C(C=C1)C2=N[N+](=[C-]N2C3=CC=CC=C3)C4=CC=CC=C4