missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(3,4-Dimethylphenyl)-3-methyl-5-pyrazolone 97.0+%, TCI America™
$96.94 - $486.52
Chemical Identifiers
| CAS | 277299-70-4 |
|---|---|
| Molecular Formula | C12H14N2O |
| Molecular Weight (g/mol) | 202.257 |
| MDL Number | MFCD09795980 |
| InChI Key | HBWBJCSXUJIDGN-UHFFFAOYSA-N |
| PubChem CID | 11735829 |
| IUPAC Name | 2-(3,4-dimethylphenyl)-5-methyl-1H-pyrazol-3-one |
| SMILES | CC1=C(C=C(C=C1)N2C(=O)C=C(N2)C)C |
Chemical Identifiers
| 277299-70-4 | |
| 202.257 | |
| HBWBJCSXUJIDGN-UHFFFAOYSA-N | |
| 2-(3,4-dimethylphenyl)-5-methyl-1H-pyrazol-3-one |
| C12H14N2O | |
| MFCD09795980 | |
| 11735829 | |
| CC1=C(C=C(C=C1)N2C(=O)C=C(N2)C)C |
Specifications
| 123°C | |
| White-Yellow | |
| Crystal-Powder at 20°C | |
| MFCD09795980 | |
| CC1=C(C=C(C=C1)N2C(=O)C=C(N2)C)C | |
| 202.257 | |
| 202.26 | |
| 1-(3,4-Dimethylphenyl)-3-methyl-5-pyrazolone |
| 277299-70-4 | |
| 1 g | |
| C12H14N2O | |
| HBWBJCSXUJIDGN-UHFFFAOYSA-N | |
| 2-(3,4-dimethylphenyl)-5-methyl-1H-pyrazol-3-one | |
| 11735829 | |
| ≥97.0% (GC,T) |
Safety and Handling
TSCA : No