missing translation for 'onlineSavingsMsg'
Learn More

1,3,4,6-Tetrathiapentalene-2,5-dione 98.0+%, TCI America™

Catalog No. p-7136624
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T11321G 1 g
T11325G 5 g
T113225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T11321G Supplier TCI America Supplier No. T11321G
Only null left

Chemical Identifiers

CAS 64394-45-2
Molecular Formula C4O2S4
Molecular Weight (g/mol) 208.282
MDL Number MFCD00010403
InChI Key XMQJUIFJLYEFQR-UHFFFAOYSA-N
PubChem CID 555056
IUPAC Name [1,3]dithiolo[4,5-d][1,3]dithiole-2,5-dione
SMILES C12=C(SC(=O)S1)SC(=O)S2

Specifications

Melting Point 178°C
Color Yellow
Quantity 1 g
Formula Weight 208.28
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3,4,6-Tetrathiapentalene-2,5-dione