accessibility menu, dialog, popup

UserName

SOSSOS

CAS RN 64394-45-2

IUPAC Name: 2H,5H-[1,3]dithiolo[4,5-d][1,3]dithiole-2,5-dione
Synonyms: [1,3]dithiolo[4,5-d][1,3]dithiole-2,5-dione
Molecular Weight (g/mol): 208.28
Molecular Formula: C4O2S4
InChi Key: XMQJUIFJLYEFQR-UHFFFAOYSA-N
SMILES: O=C1SC2=C(S1)SC(=O)S2
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results