missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3,4,6-Tetrathiapentalene-2,5-dione 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 178°C |
| Color | Yellow |
| Quantity | 5 g |
| Formula Weight | 208.28 |
| Percent Purity | ≥98.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 1,3,4,6-Tetrathiapentalene-2,5-dione |
Safety and Handling
| TSCA | No |