missing translation for 'onlineSavingsMsg'
Learn More

1,3,4,6-Tetra-O-acetyl-beta-D-mannopyranose 97.0+%, TCI America™

Catalog No. p-7136787
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T14591G 1 g
T14595G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T14591G Supplier TCI America Supplier No. T14591G

Chemical Identifiers

CAS 18968-05-3
Molecular Formula C14H20O10
Molecular Weight (g/mol) 348.304
MDL Number MFCD00012354
InChI Key SHBHJRVMGYVXKK-XVIXHAIJSA-N
PubChem CID 7269409
IUPAC Name [(2R,3R,4R,5S,6S)-3,4,6-triacetyloxy-5-hydroxyoxan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C(C(O1)OC(=O)C)O)OC(=O)C)OC(=O)C

Specifications

Melting Point 162°C
Color White-Yellow
Quantity 1 g
Formula Weight 348.30
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3,4,6-Tetra-O-acetyl-beta-D-mannopyranose
TSCA No
Recommended Storage Refrigerator