accessibility menu, dialog, popup

UserName

CAS RN 18968-05-3

IUPAC Name: [3,4,6-tris(acetyloxy)-5-hydroxyoxan-2-yl]methyl acetate
Synonyms: [3,4,6-tris(acetyloxy)-5-hydroxyoxan-2-yl]methyl acetate
Molecular Weight (g/mol): 348.3
Molecular Formula: C14H20O10
InChi Key: SHBHJRVMGYVXKK-UHFFFAOYNA-N
SMILES: CC(=O)OCC1OC(OC(C)=O)C(O)C(OC(C)=O)C1OC(C)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results