missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3,4,6,7,8-Hexahydro-4,6,6,7,8,8-hexamethylcyclopenta[g]-2-benzopyran, Analytical Standard, ≥85.0% (GC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 1222-05-5 |
| Linear Formula | C18H26O |
| MDL Number | MFCD00217003 |
| Synonym | 1,3,4,6,7,8-Hexahydro-4,6,6,7,8,8-hexamethylindeno[5,6-c]pyran; Hexamethylindanopyran |
| Quantity | 10 mg |
| Formula Weight | 258.40 |
| Percent Purity | ≥85.0% (GC) |
| Grade | Analytical Standard |