missing translation for 'onlineSavingsMsg'
Learn More

1,3,3-Trimethylindolinobenzopyrylospiran 98.0+%, TCI America™

Catalog No. p-7136231
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T03441G 1 g
T03445G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T03441G Supplier TCI America Supplier No. T03441G
Only null left

Chemical Identifiers

CAS 1485-92-3
Molecular Formula C19H19NO
Molecular Weight (g/mol) 277.37
MDL Number MFCD00059174
InChI Key CZTCZDFGLUDUQP-UHFFFAOYNA-N
PubChem CID 99764
IUPAC Name 1',3',3'-trimethyl-1',3'-dihydrospiro[chromene-2,2'-indole]
SMILES CN1C2=CC=CC=C2C(C)(C)C11OC2=CC=CC=C2C=C1

Specifications

Melting Point 92°C
Color White-Yellow
Quantity 1 g
Synonym 1′C,3′C,3′C-Trimethylspiro[1(2H)-benzopyran-2,2′C-indoline]
Formula Weight 277.37
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,3,3-Trimethylindolinobenzopyrylospiran [Photochromic Compound]