accessibility menu, dialog, popup

UserName

CAS RN 1485-92-3

H3CNCH3H3CO

CAS RN 1485-92-3

IUPAC Name: 1',3',3'-trimethyl-1',3'-dihydrospiro[chromene-2,2'-indole]
Synonyms: 1',3',3'-trimethylspiro[chromene-2,2'-indole]
Molecular Weight (g/mol): 277.37
Molecular Formula: C19H19NO
InChi Key: CZTCZDFGLUDUQP-UHFFFAOYNA-N
SMILES: CN1C2=CC=CC=C2C(C)(C)C11OC2=CC=CC=C2C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results