missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3,3-Trimethylindolinobenzopyrylospiran 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 92°C |
| Color | White-Yellow |
| Quantity | 5 g |
| Synonym | 1′,3′,3′-Trimethylspiro[1(2H)-benzopyran-2,2′-indoline] |
| Formula Weight | 277.37 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 1,3,3-Trimethylindolinobenzopyrylospiran [Photochromic Compound] |
Safety and Handling
| TSCA | Yes |