missing translation for 'onlineSavingsMsg'
Learn More

1,3,3-Trimethyl-3,4-dihydroisoquinoline 98.0+%, TCI America™

Catalog No. T30601G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T30601G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T30601G Supplier TCI America Supplier No. T30601G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 79023-51-1
Molecular Formula C12H15N
Molecular Weight (g/mol) 173.26
MDL Number MFCD00608546
InChI Key HFNOAVXTUXLYNM-UHFFFAOYSA-N
Synonym 3,4-Dihydro-1,3,3-trimethylisoquinoline
PubChem CID 594161
IUPAC Name 1,3,3-trimethyl-3,4-dihydroisoquinoline
SMILES CC1=NC(C)(C)CC2=CC=CC=C12

Specifications

Color Red
Boiling Point 125°C
Quantity 1 g
Formula Weight 173.26
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1,3,3-Trimethyl-3,4-dihydroisoquinoline