accessibility menu, dialog, popup

UserName

CAS RN 79023-51-1

IUPAC Name: 1,3,3-trimethyl-3,4-dihydroisoquinoline
Synonyms: 1,3,3-trimethyl-4H-isoquinoline
Molecular Weight (g/mol): 173.26
Molecular Formula: C12H15N
InChi Key: HFNOAVXTUXLYNM-UHFFFAOYSA-N
SMILES: CC1=NC(C)(C)CC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results