missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3,3-Trimethyl-2-methyleneindoline 96.0+%, TCI America™
$55.84 - $361.93
Chemical Identifiers
| CAS | 118-12-7 |
|---|---|
| Molecular Formula | C12H15N |
| Molecular Weight (g/mol) | 173.259 |
| MDL Number | MFCD00005813 |
| InChI Key | ZTUKGBOUHWYFGC-UHFFFAOYSA-N |
| Synonym | 1,3,3-trimethyl-2-methyleneindoline, fischer's base, fischer base, 1h-indole, 2,3-dihydro-1,3,3-trimethyl-2-methylene, fischer's methylene base, 2-methylene-1,3,3-trimethylindoline, fischers base, ccris 6608, indoline, 2-methylene-1,3,3-trimethyl, indoline, 1,3,3-trimethyl-2-methylene |
| PubChem CID | 8351 |
| IUPAC Name | 1,3,3-trimethyl-2-methylideneindole |
| SMILES | CC1(C(=C)N(C2=CC=CC=C21)C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
T087925ML
|
TCI America
T087925ML |
25 mL |
Each for $55.84
|
|
|||||
|
T0879100ML
|
TCI America
T0879100ML |
100 mL |
Each for $98.18
|
|
|||||
|
T0879500ML
|
TCI America
T0879500ML |
500 mL |
Each for $361.93
|
|
|||||
Chemical Identifiers
| 118-12-7 | |
| 173.259 | |
| ZTUKGBOUHWYFGC-UHFFFAOYSA-N | |
| 8351 | |
| CC1(C(=C)N(C2=CC=CC=C21)C)C |
| C12H15N | |
| MFCD00005813 | |
| 1,3,3-trimethyl-2-methyleneindoline, fischer's base, fischer base, 1h-indole, 2,3-dihydro-1,3,3-trimethyl-2-methylene, fischer's methylene base, 2-methylene-1,3,3-trimethylindoline, fischers base, ccris 6608, indoline, 2-methylene-1,3,3-trimethyl, indoline, 1,3,3-trimethyl-2-methylene | |
| 1,3,3-trimethyl-2-methylideneindole |
Specifications
| 118-12-7 | |
| Red | |
| C12H15N | |
| 25 mL | |
| 1,3,3-trimethyl-2-methyleneindoline, fischer's base, fischer base, 1h-indole, 2,3-dihydro-1,3,3-trimethyl-2-methylene, fischer's methylene base, 2-methylene-1,3,3-trimethylindoline, fischers base, ccris 6608, indoline, 2-methylene-1,3,3-trimethyl, indoline, 1,3,3-trimethyl-2-methylene | |
| CC1(C(=C)N(C2=CC=CC=C21)C)C | |
| 173.259 | |
| 173.26 | |
| Liquid |
| -10°C | |
| 248°C | |
| MFCD00005813 | |
| 3082 | |
| ZTUKGBOUHWYFGC-UHFFFAOYSA-N | |
| 1,3,3-trimethyl-2-methylideneindole | |
| 8351 | |
| ≥96.0% (GC) | |
| 1,3,3-Trimethyl-2-methyleneindoline |
Safety and Handling
RTECSNumber : NM1926500
TSCA : Yes