accessibility menu, dialog, popup

UserName

H3CNH2CH3CCH3

CAS RN 118-12-7

IUPAC Name: 1,3,3-trimethyl-2-methylidene-2,3-dihydro-1H-indole
Synonyms: 1,3,3-trimethyl-2-methylideneindole
Molecular Weight (g/mol): 173.26
Molecular Formula: C12H15N
InChi Key: ZTUKGBOUHWYFGC-UHFFFAOYSA-N
SMILES: CN1C(=C)C(C)(C)C2=CC=CC=C12
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)